Skip to Content
MilliporeSigma
Buffered Peptone Water in Bags according to ISO, GB, liquid, bag of 4 × 3 L, sterile

Buffered Peptone Water in Bags

Buffered Peptone Water in Bags according to ISO, GB, liquid, bag of 4 × 3 L, sterile
Synonym(s):
Buffered Peptone Water

Products

Product Category
Brand
pH Value
Physical Form
Agency
Chemical Composition
Quality Segment
Application
Microorganism Suitability
Package Size
Sterility
Technique
Showing 1-1 of 1