MilliporeSigma
vibrant-m

C18-reversed phase silica gel

vibrant-m
Synonyms:
Reversed phase silica gel 90 C18, Reversed phase silica gel C18
EC Number:

Products

Shipping
Product Category
Brand
Physical Form
Separation Technique
Quality Segment
Particle Size (µm)
Pore Size (Å)
Technique

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Showing 1-2 of 2