Skip to Content
MilliporeSigma
Cilostamide phosphodiesterase inhibitor

Cilostamide

Cilostamide phosphodiesterase inhibitor
Synonym(s):
6-[3-(N-Cyclohexyl-N-methylcarbamoyl)propoxy]quinolin-2[1H]-one, OPC 3689, Cilostamide
Empirical Formula (Hill Notation):
C20H26N2O3
CAS Number:
Molecular Weight:
342.43

Products