Citronellyl propionate ≥95%, FCC, FG

Citronellyl propionate

Citronellyl propionate ≥95%, FCC, FG
Linear Formula:
C2H5CO2CH2CH2CH(CH3)CH2CH2CH=C(CH3)2
CAS Number:
Molecular Weight:
212.33
EC Number:

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Product Number
Product Description
Pricing
Citronellyl propionate, ≥95%, FCC, FG