MilliporeSigma
Colominic acid sodium salt from Escherichia coli

Colominic acid sodium salt from Escherichia coli

Colominic acid sodium salt from Escherichia coli
Synonyms:
Poly[2,8-(N-acetylneuraminic acid sodium salt)]
Linear Formula:
(C11H16NNaO8)n
CAS Number:

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Product Number
Product Description
Pricing
Colominic acid sodium salt from Escherichia coli