Skip to Content
MilliporeSigma
Get up to 22% off for Pi Day through March 27.Save Now
CytoSoft® Elastic Moduli 16 kPa

CytoSoft®

CytoSoft® Elastic Moduli 16 kPa
EC Number:

Products

Shipping
Product Category
Brand
Physical Form
Capacity (mL)
Surface Area cm²
Wells
Package Size
Sterilization Method
Technique
Showing 1-7 of 7
Compare
Product Number
Description
Pricing