US
EN
Applications
Products
Services
Support
Home
Search Results
Diffinity RapidTip
®
2
(1)
Diffinity RapidTip
®
2
EC Number:
293-303-4
Product Comparison Guide
Use the product attributes below to configure the comparison table. (Select up to 3 total.)
Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Select Attributes
Product Number
Product Description
Pricing
D2947L
Diffinity RapidTip
®
2, for PCR Purification with Polymerase Removal, Compatible with Rainin LTS
®
pipettes
Expand
View Pricing