DMT-2′O-Methyl-rC(tac) Phosphoramidite

DMT-2′O-Methyl-rC(tac) Phosphoramidite

DMT-2′O-Methyl-rC(tac) Phosphoramidite
Synonyms:
N-[[4-(1,1-dimethylethyl)phenoxy]acetyl]-5′-O-[bis(4-methoxyphenyl)phenylmethyl]-2′-O-methyl-cytidine, 3′-[2-cyanoethyl bis(1-methylethyl)phosphoramidite], DMT-2′-O-Me-rC(tac) amidite
Empirical Formula (Hill Notation):
C52H64N5O10P
CAS Number:
Molecular Weight:
950.07

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort By: Default
Product Number
Product Description
Pricing
DMT-2′O-Methyl-rC(tac) Phosphoramidite, configured for ABI
DMT-2′O-Methyl-rC(tac) Phosphoramidite, configured for PerkinElmer, configured for Polygen
DMT-2′O-Methyl-rC(tac) Phosphoramidite