DMT-2′O-TBDMS-rC(tac) Phosphoramidite configured for ABI

DMT-2′O-TBDMS-rC(tac) Phosphoramidite

DMT-2′O-TBDMS-rC(tac) Phosphoramidite configured for ABI
Synonyms:
DMT-2′-O-TBDMS-rC(tac) amidite, N-[[4-(1,1-dimethylethyl)phenoxy]acetyl]-5′-O-[bis(4-?methoxyphenyl)phenylmethyl]-2′-O-[(1,1-dimethylethyl)dimethylsilyl]-cytidine, 3′-[2-cyanoethyl bis(1-methylethyl)phosphoramidite]
Empirical Formula (Hill Notation):
C57H76N5O10PSi
CAS Number:
Molecular Weight:
1050.30

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort By: Default
Product Number
Product Description
Pricing
DMT-2′O-TBDMS-rC(tac) Phosphoramidite, configured for ABI
DMT-2′O-TBDMS-rC(tac) Phosphoramidite, configured for (ÄKTA® and OligoPilot®)
DMT-2′O-TBDMS-rC(tac) Phosphoramidite
DMT-2′O-TBDMS-rC(tac) Phosphoramidite, configured for ABI