D-Ribulose 1,5-bisphosphate sodium salt hydrate ≥99.0% (TLC)

D-Ribulose 1,5-bisphosphate sodium salt hydrate

D-Ribulose 1,5-bisphosphate sodium salt hydrate ≥99.0% (TLC)
Synonyms:
D-erythro-2-Pentulose, 1,5-bis(dihydrogen phosphate), D-Ribulose 1,5-diphosphate, RuDP
Empirical Formula (Hill Notation):
C5H12O11P2 · xNa+ · yH2O
CAS Number:
Molecular Weight:
310.09 (anhydrous free acid basis)

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort By: Default
Product Number
Product Description
Pricing
D-Ribulose 1,5-bisphosphate sodium salt hydrate, ≥90%
D-Ribulose 1,5-bisphosphate sodium salt hydrate, ≥99.0% (TLC)

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.