Skip to Content
MilliporeSigma
Opticap® XL 4 CapsuleMedia: Durapore® 0.1 µm Chemistry: Hydrophilic Polyvinylidene Fluoride (PVDF)Inlet: 1-1/2 in. Sanitary FlangeOutlet: 9/16 in. Hose BarbMax Operating Pressure: 80 psi @ 25 °C

Durapore® 0.1 µm, Opticap® XL Capsule

Opticap® XL 4 CapsuleMedia: Durapore® 0.1 µm Chemistry: Hydrophilic Polyvinylidene Fluoride (PVDF)Inlet: 1-1/2 in. Sanitary FlangeOutlet: 9/16 in. Hose BarbMax Operating Pressure: 80 psi @ 25 °C
Synonym(s):
Durapore® 0.1 µm, Opticap® XL Capsule

Products

Product Category
Brand
Product Line
Sterilization Compatibility
Agency
Length (cm)
Length (in)
Quality Segment
Hydrophobicity
Housing Material
Support Material
Pore size (µm)
Sterility
Sterilization Method
Technique
Showing 1-20 of 31
Page 1 of 2
Compare
Product Number
Description
Pricing
Page 1 of 2