Skip to Content
MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
Durapore® Cartridge Chemistry: 0.22 µm Hydrophilic Polyvinylidene Fluoride (PVDF)Configuration: 30 in. Code 5

Durapore® 0.22 µm, Cartridge

Durapore® Cartridge Chemistry: 0.22 µm Hydrophilic Polyvinylidene Fluoride (PVDF)Configuration: 30 in. Code 5
Synonym(s):
Durapore® 0.22 µm, Cartridge

Products

Product Category
Brand
Product Line
Sterilization Compatibility
Agency
Quality Segment
Hydrophobicity
Support Material
Pore size (µm)
Sterility
Technique
Showing 1-9 of 9