Skip to Content
MilliporeSigma
Get 22% off for Pi Day through March 31.Save Now
Opticap® XL 4 CapsuleMedia: Sterile Durapore® 0.22 µm Chemistry: Hydrophilic Polyvinylidene Fluoride (PVDF)Inlet: 1-1/2 in. Sanitary FlangeOutlet: 9/16 in. Hose BarbMax Operating Pressure: 80 psi @ 25 °C

Durapore® 0.22 µm, Opticap® XL Capsule

Opticap® XL 4 CapsuleMedia: Sterile Durapore® 0.22 µm Chemistry: Hydrophilic Polyvinylidene Fluoride (PVDF)Inlet: 1-1/2 in. Sanitary FlangeOutlet: 9/16 in. Hose BarbMax Operating Pressure: 80 psi @ 25 °C
Synonym(s):
Durapore® 0.22 µm, Opticap® XL Capsule

Products

Product Category
Brand
Product Line
Sterilization Compatibility
Agency
Length (cm)
Length (in)
Quality Segment
Hydrophobicity
Housing Material
Membrane Material
Support Material
Pore size (µm)
Sterility
Sterilization Method
Technique
Showing 1-20 of 40
Page 1 of 2
Compare
Product Number
Description
Pricing
Page 1 of 2