Ethanol 96% suitable for use as excipient EMPROVE® exp Ph Eur,JP,USP

Ethanol 96%

Ethanol 96% suitable for use as excipient EMPROVE® exp Ph Eur,JP,USP
Synonyms:
Ethanol, Ethyl alcohol
Linear Formula:
CH3CH2OH
CAS Number:
Molecular Weight:
46.07

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort By: Default
Product Number
Product Description
Pricing
Ethanol 96%, EMSURE® Reag. Ph Eur
Ethanol 96%, EMPROVE® EXPERT Ph Eur,BP
Ethanol 96%, suitable for use as excipient EMPROVE® exp Ph Eur,JP,USP

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.