Ethyl 2-benzylacetoacetate 97%

Ethyl 2-benzylacetoacetate

Ethyl 2-benzylacetoacetate 97%
Linear Formula:
CH3COCH(CH2C6H5)CO2C2H5
CAS Number:
Molecular Weight:
220.26
EC Number:

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Product Number
Product Description
Pricing
Ethyl 2-benzylacetoacetate, 97%