MilliporeSigma
Flash Silica dried, pkg of 1 kg, spherical

Flash Silica

Flash Silica dried, pkg of 1 kg, spherical
Synonyms:
Silica gel
CAS Number:
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Manufacturer
Melting Point (°C)
Physical Form
Separation Technique
Particle Size (µm)
Pore Size (Å)
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing