Skip to Content
MilliporeSigma
Get up to 22% off for Pi Day through March 27.Save Now
Fmoc-Trp(Boc)-OH ≥97.0% (HPLC)

Fmoc-Trp(Boc)-OH

Fmoc-Trp(Boc)-OH ≥97.0% (HPLC)
Synonym(s):
Nα-Fmoc-N(in)-Boc-L-tryptophan, N(in)-Boc-Nα-Fmoc-L-tryptophan, Fmoc-Trp(Boc)-OH
Empirical Formula (Hill Notation):
C31H30N2O6
CAS Number:
Molecular Weight:
526.58

Products

Product Category
Brand
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Reaction Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing