Skip to Content
MilliporeSigma
Get 22% off for Pi Day through March 31.Save Now
Endoproteinase Glu-C, Excision Grade, Staphylococcus aureus

Glu-C Protease

Endoproteinase Glu-C, Excision Grade, Staphylococcus aureus
Synonym(s):
V8 Protease, Endoproteinase Glu-C, Excision Grade, Staphylococcus aureus
CAS Number:

Products

Product Category
Brand
Biological Source
Physical Form
Analyte Suitability
Quality Segment