Glycerol 85% EMPROVE® EXPERT, Ph. Eur., BP

Glycerol 85%

Glycerol 85% EMPROVE® EXPERT, Ph. Eur., BP
Synonyms:
1,2,3-Propanetriol, Glycerol solution
Linear Formula:
HOCH2CH(OH)CH2OH · aq
CAS Number:
Molecular Weight:
92.09 (anhydrous basis)

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort By: Default
Product Number
Product Description
Pricing
Glycerol 85%, for analysis EMSURE® Reag. Ph Eur
Glycerol 85%, EMPROVE® EXPERT, Ph. Eur., BP