Hexadecafluoroheptane technical grade, 85%

Hexadecafluoroheptane

Hexadecafluoroheptane technical grade, 85%
Synonyms:
Perfluoroheptane
Linear Formula:
CF3(CF2)5CF3
CAS Number:
Molecular Weight:
388.05
EC Number:

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Product Number
Product Description
Pricing
Hexadecafluoroheptane, technical grade, 85%