vibrant-m

Hexanes, 98.5%

vibrant-m
Synonyms:
Hexanes
Empirical Formula (Hill Notation):
C6H14
CAS Number:
Molecular Weight:
86.18

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort By: Default
Product Number
Product Description
Pricing
Hexanes, 98.5%, OmniSolv®
Hexanes, 98.5%, for GC analyses, Non-UV OmniSolv®
Hexanes, 98.5%, OmniSolv® HR-GC