Hydrogen peroxide 35% EMPLURA®

Hydrogen peroxide 35%

Hydrogen peroxide 35% EMPLURA®
Synonyms:
Hydrogen peroxide solution
Empirical Formula (Hill Notation):
H2O2
CAS Number:
Molecular Weight:
34.01

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort By: Default
Product Number
Product Description
Pricing
Hydrogen peroxide 35%
Hydrogen peroxide 35%, EMPLURA®