Indole-6-carboxylic acid 97%

Indole-6-carboxylic acid

Indole-6-carboxylic acid 97%
Synonyms:
6-Carboxyindole
Empirical Formula (Hill Notation):
C9H7NO2
CAS Number:
Molecular Weight:
161.16

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Product Number
Product Description
Pricing
Indole-6-carboxylic acid, 97%