Ipriflavone analytical standard

Ipriflavone

Ipriflavone analytical standard
Synonyms:
7-Isopropoxy-3-phenyl-4H-1-benzopyran-4-one, Ipriflavone
Empirical Formula (Hill Notation):
C18H16O3
CAS Number:
Molecular Weight:
280.32

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Product Number
Product Description
Pricing
Ipriflavone, analytical standard