Skip to Content
MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
IWR-1 ≥98% (HPLC)

IWR-1

IWR-1 ≥98% (HPLC)
Synonym(s):
4-(1,3,3a,4,7,7a-Hexahydro-1,3-dioxo-4,7-methano-2H-isoindol-2-yl)-N-8-quinolinyl-Benzamide, IWR1, InSolution Wnt Antagonist I, IWR-1-endo
Empirical Formula (Hill Notation):
C25H19N3O3
CAS Number:
Molecular Weight:
409.44

Products

Shipping
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Research Area
Quality Segment
Showing 1-2 of 2
Compare
Product Number
Description
Pricing