Luperox® TBH70X, tert-Butyl hydroperoxide solution 70 wt. % in H2O

Luperox® TBH70X, tert-Butyl hydroperoxide solution

Luperox® TBH70X, tert-Butyl hydroperoxide solution 70 wt. % in H2O
Synonyms:
tert-Butyl hydroperoxide solution, 1,1-Dimethylethyl hydroperoxide, TBHP, 2-Hydroperoxy-2-methylpropane
Linear Formula:
(CH3)3COOH
CAS Number:
Molecular Weight:
90.12

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort By: Default
Product Number
Product Description
Pricing
Luperox® TBH70X, tert-Butyl hydroperoxide solution, 70 wt. % in H2O

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.