US
EN
Applications
Products
Services
Support
Home
Search Results
Magnesium sulfate solution
(2)
Magnesium sulfate solution
Empirical Formula (Hill Notation):
MgSO
4
CAS Number:
7487-88-9
Molecular Weight:
120.37
Product Comparison Guide
Use the product attributes below to configure the comparison table. (Select up to 3 total.)
Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Select Attributes
Product Number
Product Description
Pricing
M3409
Magnesium sulfate solution, for molecular biology, 1.00 M±0.04 M
Expand
View Pricing
83266
Magnesium sulfate solution, BioUltra, for molecular biology
Expand
View Pricing