Products
US
EN
Products
Applications
Products
Services
Support
Home
Search Results
m
-Anisic acid
(1)
m
-Anisic acid
Synonyms:
m
-Methylsalicylic acid,
m
-Anisic acid, 3-Methoxybenzoic acid
Linear Formula:
CH
3
OC
6
H
4
CO
2
H
CAS Number:
586-38-9
Molecular Weight:
152.15
EC Number:
209-574-9
Product Comparison Guide
Use the product attributes below to configure the comparison table. (Select up to 3 total.)
Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Select Attributes
Product Number
Product Description
Pricing
W394440
m
-Anisic acid, ≥98%
Expand
View Pricing