Skip to Content
MilliporeSigma
Mdivi-1 ≥98% (HPLC), powder

Mdivi-1

Mdivi-1 ≥98% (HPLC), powder
Synonym(s):
3-(2,4-Dichloro-5-methoxyphenyl)-2,3-dihydro-2-thioxo-4(1H)-quinazolinone, 3-(2,4-Dichloro-5-methoxyphenyl)-2-sulfanyl-4(3H)-quinazolinone, Mitochondrial Division Inhibitor, mdivi-1
Empirical Formula (Hill Notation):
C15H10Cl2N2O2S
CAS Number:
Molecular Weight:
353.22

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Research Area
Quality Segment