Skip to Content
MilliporeSigma
Get up to 22% off for Pi Day through March 27.Save Now
Medroxyprogesterone Acetate Related Compound A certified reference material, pharmaceutical secondary standard

Medroxyprogesterone acetate Related Compound A

Medroxyprogesterone Acetate Related Compound A certified reference material, pharmaceutical secondary standard
Synonym(s):
4,5-β-Dihydromedroxyprogesterone acetate, 6alpha-Methyl-3,20-dioxopregnan-17-yl acetate
Empirical Formula (Hill Notation):
C24H36O4
Molecular Weight:
388.54

Products

Shipping
Product Category
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade