MilliporeSigma
Menthalactone ≥99%

Menthalactone

Menthalactone ≥99%
Empirical Formula (Hill Notation):
C10H14O2
CAS Number:
Molecular Weight:
166.22
EC Number:

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Product Number
Product Description
Pricing
Menthalactone, ≥99%