Products
US
EN
Products
Applications
Products
Services
Support
Home
Search Results
Methyl 2-hexynoate
(1)
Methyl 2-hexynoate
Linear Formula:
CH
3
CH
2
CH
2
C≡CCO
2
CH
3
CAS Number:
18937-79-6
Molecular Weight:
126.15
EC Number:
242-690-8
Product Comparison Guide
Use the product attributes below to configure the comparison table. (Select up to 3 total.)
Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Select Attributes
Product Number
Product Description
Pricing
649090
Methyl 2-hexynoate, 98%
Expand
View Pricing