Skip to Content
MilliporeSigma
Methylone hydrochloride 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Methylone hydrochloride

Methylone hydrochloride 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Synonym(s):
2-Methylamino-3,4-methylene-dioxy-propiophenone hydrochloride, Methylenedioxymethcathinone hydrochloride, Methylone hydrochloride solution
Empirical Formula (Hill Notation):
C11H13NO3 · HCl
CAS Number:
Molecular Weight:
243.69
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Purity
Format
Research Area
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing