
Millipore Express® SHF 0.2µm, Opticap® XLT 10 Capsule

Synonym(s):
Millipore Express® SHF, Opticap® XLT Capsule
Products
Product Category
Brand
Product Line
Sterilization Compatibility
Agency
Length (cm)
Length (in)
Quality Segment
Hydrophobicity
Housing Material
Membrane Material
Support Material
Pore size (µm)
Sterility
Sterilization Method
Technique
Showing 1-15 of 15
Compare
Product Number
Description
Pricing
Millipore Express® SHF 0.2µm, Opticap® XLT 10 Capsule, Fittings 1-1/2 in. TC- 1 in. HB, Autoclavable