
Millipore Express® SHR Cartridge

Synonym(s):
Millipore Express® SHR Cartridge Filter 10 in. 0.1 µm Code 7 Silicone, Millipore Express® SHR Cartridge
Products
Product Category
Product Line
Quality Segment
Pore size (µm)
Sterility
Technique
Showing 1-2 of 2