MilliporeSigma
Ammonia solution 2.0 M in isopropanol

Ammonia solution

Ammonia solution 2.0 M in isopropanol
Linear Formula:
NH3
CAS Number:
Molecular Weight:
17.03

Products

Shipping
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Quality Segment
Application
Technique