
Monoclonal Anti-ALDH2 antibody produced in mouse

Synonym(s):
Anti-ALDHE2, Anti-ALDHI, Anti-ALDM, Anti-MGC1806, Anti-aldehyde dehydrogenase 2 family (mitochondrial)
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Isotype
Physical Form
Antibody Form
Research Area
Conjugate
Post Translational Modification
Species Reactivity
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing