Skip to Content
MilliporeSigma
Get 22% off for Pi Day through March 31.Save Now
Monoclonal Anti-CRP antibody produced in mouse clone C7

Monoclonal Anti-CRP antibody produced in mouse

Monoclonal Anti-CRP antibody produced in mouse clone C7

Products

Product Category
Brand
Biological Source
Clonality
Gene Alias
Physical Form
Antibody Form
Conjugate
Post Translational Modification
Technique
Showing 1-2 of 2