
MONOCLONAL ANTI-MOUSE IRGM1 (C-TERMINAL) antibody produced in mouse

Synonym(s):
Ifi1, Iigp3, Immunity-related GTPase family M protein 1, Irgm, Irgm1
Products
Product Category
Brand
Biological Source
Clonality
Physical Form
Conjugate
Quality Segment
Species Reactivity
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing