N-Acetylanthranilic acid 99%

N-Acetylanthranilic acid

N-Acetylanthranilic acid 99%
Synonyms:
2-Acetamidobenzoic acid
Linear Formula:
(CH3CONH)C6H4CO2H
CAS Number:
Molecular Weight:
179.17
EC Number:

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Product Number
Product Description
Pricing
N-Acetylanthranilic acid, 99%