MilliporeSigma
NanoFabTx™ microfluidic - tubing 250um FEP with ferrules, 10m

NanoFabTx microfluidic - tubing

NanoFabTx™ microfluidic - tubing 250um FEP with ferrules, 10m

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Product Number
Product Description
Pricing
NanoFabTx microfluidic - tubing, 100μm FEP with ferrules, 10m
NanoFabTx microfluidic - tubing, 250um FEP with ferrules, 10m