n-Hexane 95% Meets ACS Specifications GR ACS

n-Hexane 95%

n-Hexane 95% Meets ACS Specifications GR ACS
Synonyms:
n-Hexane, Hexane
Linear Formula:
CH3(CH2)4CH3
CAS Number:
Molecular Weight:
86.18

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort By: Default
Product Number
Product Description
Pricing
n-Hexane 95%, OmniSolv®
n-Hexane 95%, HPLC, Meets Reagent Specifications for testing USP/NF monographs
n-Hexane 95%, Meets ACS Specifications GR ACS