N,N-Bis[2-(p-tolylsulfonyloxy)ethyl]-p-toluenesulfonamide 97%

N,N-Bis[2-(p-tolylsulfonyloxy)ethyl]-p-toluenesulfonamide

N,N-Bis[2-(p-tolylsulfonyloxy)ethyl]-p-toluenesulfonamide 97%
Synonyms:
N,N-Bis[2-(tosyloxy)ethyl]-p-toluenesulfonamide, N,O,O′-Tritosyldiethanolamine
Linear Formula:
CH3C6H4SO2N(CH2CH2OSO2C6H4CH3)2
CAS Number:
Molecular Weight:
567.69
EC Number:

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort By: Default
Product Number
Product Description
Pricing
N,N-Bis[2-(p-tolylsulfonyloxy)ethyl]-p-toluenesulfonamide, 97%

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.