US
EN
Applications
Products
Services
Support
Home
Search Results
n-Octane
(2)
n-Octane
Synonyms:
n-Octane
Linear Formula:
CH
3
(CH
2
)
6
CH
3
CAS Number:
111-65-9
Molecular Weight:
114.23
Product Comparison Guide
Use the product attributes below to configure the comparison table. (Select up to 3 total.)
Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Select Attributes
Product Number
Product Description
Pricing
8.06910
n-Octane, for synthesis
Expand
View Pricing
1.09716
n-Octane, reference substance for gas chromatography
Expand
View Pricing