US
EN
Applications
Products
Services
Support
Home
Search Results
Octadecanamide
(1)
Octadecanamide
Synonyms:
Amide C
18
, Stearamide
Linear Formula:
CH
3
(CH
2
)
16
CONH
2
CAS Number:
124-26-5
Molecular Weight:
283.49
EC Number:
204-693-2
Product Comparison Guide
Use the product attributes below to configure the comparison table. (Select up to 3 total.)
Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Select Attributes
Product Number
Product Description
Pricing
O601
Octadecanamide, technical grade, 85%
Expand
View Pricing