Okadaic acid from Prorocentrum concavum 92-100% (HPLC)

Okadaic acid from Prorocentrum concavum

Okadaic acid from Prorocentrum concavum 92-100% (HPLC)
Synonyms:
OA
Empirical Formula (Hill Notation):
C44H68O13
CAS Number:
Molecular Weight:
805.00

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Product Number
Product Description
Pricing
Okadaic acid from Prorocentrum concavum, 92-100% (HPLC)