Products
US
EN
Products
Applications
Products
Services
Support
Home
Search Results
OmniPur
®
EDTA
(2)
OmniPur
®
EDTA
Linear Formula:
(HOOCCH
2
)
2
NCH
2
CH
2
N(CH
2
COOH)
2
CAS Number:
60-00-4
Product Comparison Guide
Use the product attributes below to configure the comparison table. (Select up to 3 total.)
Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Select Attributes
Product Number
Product Description
Pricing
4005-OP
OmniPur
®
EDTA
Expand
View Pricing
4008-M
OmniPur
®
EDTA
Expand
View Pricing