MilliporeSigma
Parteck® MXP (Polyvinyl alcohol) EMPROVE® ESSENTIAL, Ph. Eur., ChP, JPE, USP

Parteck® MXP (Polyvinyl alcohol)

Parteck® MXP (Polyvinyl alcohol) EMPROVE® ESSENTIAL, Ph. Eur., ChP, JPE, USP
Synonyms:
Parteck® Polyvinyl alcohol
Linear Formula:
(C2H4O)n
CAS Number:

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Product Number
Product Description
Pricing
Parteck® MXP (Polyvinyl alcohol), EMPROVE® ESSENTIAL, Ph. Eur., ChP, JPE, USP