Skip to Content
US
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
PhosphoDetect Anti-PDH-E1α (pSer²³²) Rabbit pAb
Products
Product Category
antibodies (2)
primary antibodies (2)
Brand
Sigma-Aldrich (3)
Biological Source
Clonality
Physical Form
Antibody Form
Quality Segment
Post Translational Modification
Species Reactivity
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
AP1063
PhosphoDetect Anti-PDH-E1α (pSer²³²) Rabbit pAb,
liquid, Calbiochem
®
Expand
View Pricing
AP1064
PhosphoDetect Anti-PDH-E1α (pSer²³²) Rabbit pAb,
liquid, Calbiochem
®
Expand
View Pricing
AP1062
PhosphoDetect Anti-PDH-E1α (pSer²³²) Rabbit pAb,
Expand
View Pricing