Skip to Content
MilliporeSigma
Get 22% off for Pi Day through March 31.Save Now
vibrant-m

Porcine Serum

vibrant-m

Products

Product Category
Brand
Biological Source
Chemical Composition
Quality Segment
Geographic Origin
Sterilization Method
Technique
Showing 1-1 of 1